missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2,2,3-Trimethylnorborn-5-yl)cyclohexyl acrylate

Manufacturer:  Alfa Aesar™ 4653914

 View more versions of this product

Catalog No. AA4653914



Specifications

Specifications

3- (2,2,3-Trimethylnorborn-5-yl)cyclohexyl acrylate
Brown
903876-45-9
MFCD22380703
CDBRNGRSVNBVLJ-UHFFFAOYNA-N
290.45
>149°C (300°F)
25g
C19H30O2
3-5,5,6-trimethylbicyclo 2.2.1 heptan-2-yl cyclohexyl prop-2-enoate
290.45
Liquid

Chemical Identifiers

903876-45-9
290.45
CDBRNGRSVNBVLJ-UHFFFAOYNA-N
C19H30O2
MFCD22380703
3-5,5,6-trimethylbicyclo 2.2.1 heptan-2-yl cyclohexyl prop-2-enoate
Safety and Handling

Safety and Handling

GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Recommended Storage : 2°C to 8°C

SDS Documents