Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(1H-Tetrazol-5-yl)benzyl alcohol, 96%

$80.61 - $200.41

Chemical Identifiers

Chemical Name or Material 3-(1H-Tetrazol-5-yl)benzyl alcohol
CAS 179057-20-6
Molecular Formula C8H8N4O
Formula Weight 176.18
MDL Number MFCD09864536
Synonym 5-(3-Hydroxymethylphenyl)-1H-tetrazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2698503

  SDS
alfa aesar™
H2698503
1g Each for $200.41

AAH26985MD

  SDS
alfa aesar™
H26985MD
250mg Each for $80.61
Description & Specifications

Specifications

Formula Weight 176.18
Synonym 5-(3-Hydroxymethylphenyl)-1H-tetrazole
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 208° to 210°C
Health Hazard 1 Warning
Chemical Name or Material 3-(1H-Tetrazol-5-yl)benzyl alcohol
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 179057-20-6
Molecular Formula C8H8N4O
Assay 96%
MDL Number MFCD09864536
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.