Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(1H-Tetrazol-5-yl)aniline, 96%

$79.73 - $79.73

Chemical Identifiers

Chemical Name or Material 3-(1H-Tetrazol-5-yl)aniline
CAS 73732-51-1
Molecular Formula C7H7N5
Formula Weight 161.17
MDL Number MFCD01219561
Synonym 5-(3-Aminophenyl)tetrazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2250209

  SDS
alfa aesar™
B2250209
10g Each for $79.73
Description & Specifications

Specifications

Formula Weight 161.17
Chemical Name or Material 3-(1H-Tetrazol-5-yl)aniline
Assay 96%
Molecular Formula C7H7N5
CAS 73732-51-1
Synonym 5-(3-Aminophenyl)tetrazole
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 202° to 205°C
MDL Number MFCD01219561
RTECS Number BY9276000
Quantity 10g

Applications
Used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.