Please login to your online account to display your discounted pricing

3-(1H-Pyrrol-1-yl)thiophene-2-carboxylic acid, 97%


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC22101DA

 View more versions of this product

Catalog No. CC22101DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 193.23
Quantity 1g
CAS 74772-17-1
Packaging Amber Glass Bottle
Assay Percent Range 97%
Chemical Name or Material 3-(1H-Pyrrol-1-yl)thiophene-2-carboxylic acid

Specifications

Chemical Name or Material 3-(1H-Pyrrol-1-yl)thiophene-2-carboxylic acid
Assay Percent Range 97%
Formula Weight 193.23