Please login to your online account to display your discounted pricing

3-(1H-Pyrrol-1-yl)thiophene-2-carboxylic acid, 97%

Chemical Identifiers

Chemical Name or Material 3-(1H-Pyrrol-1-yl)thiophene-2-carboxylic acid
Assay Percent Range 97%
Formula Weight 193.23
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC22101DA

  SDS
fisher scientific company
CC22101DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 193.23
Quantity 1g
CAS 74772-17-1
Packaging Amber Glass Bottle
Assay Percent Range 97%
Chemical Name or Material 3-(1H-Pyrrol-1-yl)thiophene-2-carboxylic acid