Please login to your online account to display your discounted pricing

3-(1H-Pyrrol-1-yl)propanenitrile, 97%, Maybridge


Quantity 10g

Manufacturer: fisher scientific company  CD01949EA

 View more versions of this product

Catalog No. CD01949EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 120.15
Quantity 10g
CAS 43036-06-2
Assay Percent Range 97%
Molecular Formula C7H8N2
Synonym 1-Pyrrolepropionitrile
Chemical Name or Material 3-(1H-Pyrrol-1-yl)propanenitrile

Specifications

Chemical Name or Material 3-(1H-Pyrrol-1-yl)propanenitrile
Assay Percent Range 97%
Molecular Formula C7H8N2
Formula Weight 120.15
Synonym 1-Pyrrolepropionitrile