Please login to your online account to display your discounted pricing

3-(1H-Pyrrol-1-yl)benzene-1-carbothioamide, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 3-(1H-pyrrol-1-yl)benzene-1-carbothioamide
CAS 175276-79-6
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H10N2S
Formula Weight 202.28
MDL Number MFCD00052522
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

SPB01074DA

  SDS
fisher scientific company
SPB01074DA
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SPB01074EA

  SDS
fisher scientific company
SPB01074EA
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 202.28
Quantity 1g
Assay 97%
CAS Min % 97.0
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Assay Percent Range 97%
CAS 175276-79-6
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 3-(1H-pyrrol-1-yl)benzene-1-carbothioamide
MDL Number MFCD00052522
Molecular Formula C11H10N2S
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.