missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(1H-Pyrrol-1-yl)benzaldehyde, 95+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC21904EA
Specifications
3-(1H-Pyrrol-1-yl)benzaldehyde | |
95% | |
C11H9NO | |
10g | |
PALTUANHIBXQMX-UHFFFAOYSA-N | |
3-pyrrol-1-ylbenzaldehyde | |
2776528 | |
95% |
129747-77-9 | |
Amber Glass Bottle | |
MFCD03086140 | |
3-1h-pyrrol-1-yl benzaldehyde, 3-pyrrol-1-yl-benzaldehyde, 3-pyrrol-1-yl benzaldehyde, benzaldehyde,3-1h-pyrrol-1-yl, benzaldehyde, 3-1h-pyrrol-1-yl, acmc-20airb, 3-pyrrolylbenzaldehyde, 3-1-pyrrolyl benzaldehyde | |
O=CC1=CC(=CC=C1)N1C=CC=C1 | |
171.20 | |
171.2 |
Chemical Identifiers
129747-77-9 | |
171.20 | |
PALTUANHIBXQMX-UHFFFAOYSA-N | |
2776528 | |
O=CC1=CC(=CC=C1)N1C=CC=C1 |
C11H9NO | |
MFCD03086140 | |
3-1h-pyrrol-1-yl benzaldehyde, 3-pyrrol-1-yl-benzaldehyde, 3-pyrrol-1-yl benzaldehyde, benzaldehyde,3-1h-pyrrol-1-yl, benzaldehyde, 3-1h-pyrrol-1-yl, acmc-20airb, 3-pyrrolylbenzaldehyde, 3-1-pyrrolyl benzaldehyde | |
3-pyrrol-1-ylbenzaldehyde |