Please login to your online account to display your discounted pricing

3-(1H-Pyrrol-1-yl)benzaldehyde, 95+%, Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  CC21904CB

 View more versions of this product

Catalog No. CC21904CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 171.2
Quantity 250mg
Assay 95%
Chemical Name or Material 3-(1H-Pyrrol-1-yl)benzaldehyde
Assay Percent Range 95%
Molecular Formula C11H9NO
CAS 129747-77-9

Specifications

Chemical Name or Material 3-(1H-Pyrrol-1-yl)benzaldehyde
Assay 95%
Assay Percent Range 95%
Molecular Formula C11H9NO
Formula Weight 171.2