missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(1H-Pyrrol-1-yl)aniline, 97%, Maybridge™

Manufacturer:  Fisher Scientific Company CC21914EA

 View more versions of this product

Catalog No. CC21914EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

3-(1H-pyrrol-1-yl)aniline
10g
96.5
97%
MFCD03407309
PJGDCPOPSNUYHC-UHFFFAOYSA-N
3-pyrrol-1-ylaniline
2760546
97%
Amber Glass Bottle
89353-42-4
100.0
C10H10N2
1-3-aminophenyl pyrrole, 3-1-pyrrolyl aniline, 3-1h-pyrrol-1-yl aniline, 3-pyrrol-1-yl aniline, 3-pyrrol-1-yl-aniline, 3-pyrrol-1-yl-phenylamine, 3-pyrrolylphenylamine, benzenamine, 3-1h-pyrrol-1-yl
C1=CN(C=C1)C2=CC(=CC=C2)N
158.204
158.2

Chemical Identifiers

89353-42-4
158.204
PJGDCPOPSNUYHC-UHFFFAOYSA-N
2760546
C1=CN(C=C1)C2=CC(=CC=C2)N
C10H10N2
MFCD03407309
1-3-aminophenyl pyrrole, 3-1-pyrrolyl aniline, 3-1h-pyrrol-1-yl aniline, 3-pyrrol-1-yl aniline, 3-pyrrol-1-yl-aniline, 3-pyrrol-1-yl-phenylamine, 3-pyrrolylphenylamine, benzenamine, 3-1h-pyrrol-1-yl
3-pyrrol-1-ylaniline
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents