Please login to your online account to display your discounted pricing

3-(1H-Pyrazol-3-yl)benzoic acid, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC31701DA

 View more versions of this product

Catalog No. CC31701DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 188.19
Quantity 1g
Assay 97%
CAS 850375-11-0
Assay Percent Range 97%
Chemical Name or Material 3-(1H-Pyrazol-3-yl)benzoic acid
Packaging Amber Glass Bottle
Molecular Formula C10H8N2O2

Specifications

Chemical Name or Material 3-(1H-Pyrazol-3-yl)benzoic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H8N2O2
Formula Weight 188.19