Please login to your online account to display your discounted pricing

3-(1H-Imidazol-1-ylmethyl)benzoic acid hydrochloride, Tech., Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  CC43251CB

 View more versions of this product

Catalog No. CC43251CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 238.67
Purity Grade Technical Grade
Quantity 250mg
Assay 90%
CAS Max % 100.0
Chemical Name or Material 3-(1H-Imidazol-1-ylmethyl)benzoic acid hydrochloride
MDL Number MFCD08690248
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 89.5
Assay Percent Range 90%
Health Hazard 1 GHS Signal Word: Warning
Molecular Formula C11H10N2O2·ClH
CAS 218131-32-9

Specifications

Chemical Name or Material 3-(1H-Imidazol-1-ylmethyl)benzoic acid hydrochloride
CAS 218131-32-9
CAS Min % 89.5
CAS Max % 100.0
Purity Grade Technical Grade
Assay 90%
Assay Percent Range 90%
Molecular Formula C11H10N2O2·ClH
Formula Weight 238.67
MDL Number MFCD08690248
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents