Please login to your online account to display your discounted pricing

3-(1H-Imidazol-1-ylmethyl)aniline, 97%, Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  CC43214CB

 View more versions of this product

Catalog No. CC43214CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 173.22
Quantity 250mg
Assay 97%
CAS 120107-85-9
Assay Percent Range 97%
Molecular Formula C10H11N3
Chemical Name or Material 3-(1H-Imidazol-1-ylmethyl)aniline

Specifications

Chemical Name or Material 3-(1H-Imidazol-1-ylmethyl)aniline
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H11N3
Formula Weight 173.22