missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(1H-Imidazol-1-yl)benzoic acid, ≥97%, Maybridge

Chemical Identifiers

CAS 108035-47-8
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C10H8N2O2
Formula Weight 188.19
MDL Number MFCD06659077
Chemical Name or Material 3-(1H-imidazol-1-yl)benzoic acid
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC32601CB SDS Fisher Scientific Company
CC32601CB
Amber glass bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC32601DA SDS Fisher Scientific Company
CC32601DA
Amber glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC32601DE SDS Fisher Scientific Company
CC32601DE
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

108035-47-8
100.0
C10H8N2O2
MFCD06659077
97%
96.5
97%
188.19
3-(1H-imidazol-1-yl)benzoic acid

Specifications

188.19
100.0
C10H8N2O2
Amber glass bottle
108035-47-8
97%
250mg
97%
96.5
MFCD06659077
3-(1H-imidazol-1-yl)benzoic acid

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.