missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(1H-Imidazol-1-yl)benzoic acid, ≥97%, Maybridge

Manufacturer:  Fisher Scientific Company CC32601DE

 View more versions of this product

Catalog No. CC32601DE

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

188.19
97%
C10H8N2O2
MFCD06659077
3-(1H-imidazol-1-yl)benzoic acid
5g
100.0
108035-47-8
97%
96.5

Chemical Identifiers

108035-47-8
100.0
C10H8N2O2
MFCD06659077
97%
96.5
97%
188.19
3-(1H-imidazol-1-yl)benzoic acid

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.