Please login to your online account to display your discounted pricing

3-(1h-1,2,4-Triazol-1-ylmethyl)benzaldehyde, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC45404CB

 View more versions of this product

Catalog No. CC45404CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 187.2
Quantity 250mg
Assay 97%
Chemical Name or Material 3-(1H-1, 2, 4-Triazol-1-ylmethyl)benzaldehyde
Assay Percent Range 97%
Molecular Formula C10H9N3O
CAS 876316-30-2
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 3-(1H-1, 2, 4-Triazol-1-ylmethyl)benzaldehyde
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H9N3O
Formula Weight 187.2