Please login to your online account to display your discounted pricing

3-(1h-1,2,4-Triazol-1-yl)aniline, 90%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC38414DA

 View more versions of this product

Catalog No. CC38414DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 160.18
Quantity 1g
Assay 90%
Chemical Name or Material 3-(1H-1, 2, 4-Triazol-1-yl)aniline
Packaging Amber Glass Bottle
Assay Percent Range 90%
Molecular Formula C8H8N4
CAS 176032-78-3

Specifications

Chemical Name or Material 3-(1H-1, 2, 4-Triazol-1-yl)aniline
Assay 90%
Assay Percent Range 90%
Molecular Formula C8H8N4
Formula Weight 160.18