missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(1-Pyrrolylmethyl)pyridine 97.0+%, TCI America™
$228.94 - $819.54
Chemical Identifiers
CAS | 80866-95-1 |
---|---|
Molecular Formula | C10H10N2 |
Molecular Weight (g/mol) | 158.20 |
MDL Number | MFCD00006405 |
InChI Key | PJVWPGGKILHMKW-UHFFFAOYSA-N |
PubChem CID | 697607 |
IUPAC Name | 3-[(1H-pyrrol-1-yl)methyl]pyridine |
SMILES | C(N1C=CC=C1)C1=CC=CN=C1 |
Chemical Identifiers
80866-95-1 | |
158.20 | |
PJVWPGGKILHMKW-UHFFFAOYSA-N | |
3-[(1H-pyrrol-1-yl)methyl]pyridine |
C10H10N2 | |
MFCD00006405 | |
697607 | |
C(N1C=CC=C1)C1=CC=CN=C1 |
Specifications
80866-95-1 | |
C10H10N2 | |
1 g | |
C(N1C=CC=C1)C1=CC=CN=C1 | |
158.20 | |
158.20 | |
Crystalline Powder |
60°C | |
MFCD00006405 | |
PJVWPGGKILHMKW-UHFFFAOYSA-N | |
3-[(1H-pyrrol-1-yl)methyl]pyridine | |
697607 | |
≥97.0% (GC,T) | |
3-(1-Pyrrolylmethyl)pyridine |
Safety and Handling
TSCA : No