Please login to your online account to display your discounted pricing

[3-(1-Pyrrolidinylmethyl)phenyl]methanamine, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material [3-(1-Pyrrolidinylmethyl)phenyl]methanamine
Assay 90%
Assay Percent Range 90%
Molecular Formula C12H18N2
Formula Weight 190.29
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC55713CB

  SDS
fisher scientific company
CC55713CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC55713EA

 
fisher scientific company
CC55713EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC55713DA

  SDS
fisher scientific company
CC55713DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 190.29
Quantity 250mg
Assay 90%
CAS 91271-78-2
Assay Percent Range 90%
Packaging Amber Glass Bottle
Chemical Name or Material [3-(1-Pyrrolidinylmethyl)phenyl]methanamine
Molecular Formula C12H18N2