Please login to your online account to display your discounted pricing

[3-(1-Pyrrolidinylmethyl)phenyl]methanamine, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC55713EA

 View more versions of this product

Catalog No. CC55713EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 190.29
Quantity 10g
Assay 90%
Chemical Name or Material [3-(1-Pyrrolidinylmethyl)phenyl]methanamine
Packaging Amber Glass Bottle
Assay Percent Range 90%
Molecular Formula C12H18N2
CAS 91271-78-2

Specifications

Chemical Name or Material [3-(1-Pyrrolidinylmethyl)phenyl]methanamine
Assay 90%
Assay Percent Range 90%
Molecular Formula C12H18N2
Formula Weight 190.29