missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(1-Pyrrolidinylmethyl)indole, 95%

Manufacturer:  Alfa Aesar™ H63840MD

 View more versions of this product

Catalog No. AAH63840MD



Description

Description

Applications
Reatant in the synthesis 6-substituted 7-methyl-1,4-dioxa-7-azaspiro[4.5]decanes.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids, bases.
Specifications

Specifications

3-(1-Pyrrolidinylmethyl)indole
250mg
C13H16N2
477183
ZIIMZMSKBKAABD-UHFFFAOYSA-N
3-(pyrrolidin-1-ylmethyl)-1H-indole
21478
Solid
White
5379-94-2
MFCD00450050
3-1-pyrrolidinylmethyl-1h-indole, n-skatylpyrrolidine, 3-pyrrolidinomethyl indole, 1h-indole, 3-1-pyrrolidinylmethyl, 3-pyrrolidin-1-ylmethyl-1h-indole, 3-pyrrolidin-1-ylmethyl indole, 5-22-10-00028 beilstein handbook reference, 3-pyrrolizinomethyl-1h-indole, 3-1-pyrrolidinylmethyl indole
C1CCN(C1)CC2=CNC3=CC=CC=C32
200.285
200.28
95%

Chemical Identifiers

5379-94-2
200.285
ZIIMZMSKBKAABD-UHFFFAOYSA-N
21478
C1CCN(C1)CC2=CNC3=CC=CC=C32
C13H16N2
MFCD00450050
3-1-pyrrolidinylmethyl-1h-indole, n-skatylpyrrolidine, 3-pyrrolidinomethyl indole, 1h-indole, 3-1-pyrrolidinylmethyl, 3-pyrrolidin-1-ylmethyl-1h-indole, 3-pyrrolidin-1-ylmethyl indole, 5-22-10-00028 beilstein handbook reference, 3-pyrrolizinomethyl-1h-indole, 3-1-pyrrolidinylmethyl indole
3-(pyrrolidin-1-ylmethyl)-1H-indole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : NM1871500

SDS Documents