Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(1-Pyrrolidinylcarbonyl)benzeneboronic acid pinacol ester, 97%

$86.34 - $290.81

Chemical Identifiers

Chemical Name or Material 3-(1-Pyrrolidinylcarbonyl)benzeneboronic acid pinacol ester
CAS 1073353-61-3
Molecular Formula C17H24BNO3
Formula Weight 301.19
MDL Number MFCD09027086
Synonym 3-(Pyrrolidine-1-carbonyl)phenylboronic acid pinacol ester; 1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]pyrrolidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2880103

  SDS
alfa aesar™
H2880103
1g Each for $86.34

AAH2880106

  SDS
alfa aesar™
H2880106
5g Each for $290.81
Description & Specifications

Specifications

Formula Weight 301.19
Chemical Name or Material 3-(1-Pyrrolidinylcarbonyl)benzeneboronic acid pinacol ester
Assay 97%
Molecular Formula C17H24BNO3
MDL Number MFCD09027086
CAS 1073353-61-3
Synonym 3-(Pyrrolidine-1-carbonyl)phenylboronic acid pinacol ester; 1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]pyrrolidine
Melting Point 96° to 98°C
Quantity 1g