Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(1-Piperidinylmethyl)phenol, 98%

$54.65 - $463.32

Chemical Identifiers

Chemical Name or Material 3-(1-Piperidinylmethyl)phenol
CAS 73279-04-6
Molecular Formula C12H17NO
Formula Weight 191.27
MDL Number MFCD00800250
Synonym 1-(3-Hydroxybenzyl)piperidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2763903

 
alfa aesar™
H2763903
1g Each for $54.65

AAH2763906

 
alfa aesar™
H2763906
5g Each for $157.00

AAH2763914

 
alfa aesar™
H2763914
25g Each for $463.32
Description & Specifications

Specifications

Formula Weight 191.27
Chemical Name or Material 3-(1-Piperidinylmethyl)phenol
Assay 98%
Molecular Formula C12H17NO
MDL Number MFCD00800250
CAS 73279-04-6
Synonym 1-(3-Hydroxybenzyl)piperidine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 138° to 142°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.