Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(1-Piperidinylmethyl)indole, 95%

Chemical Identifiers

Chemical Name or Material 3-(1-Piperidinylmethyl)indole
CAS 5355-42-0
Formula Weight 214.3
MDL Number MFCD00458412
Beilstein 176547
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6325603

  SDS
alfa aesar™
H6325603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6325606

  SDS
alfa aesar™
H6325606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63256MD

  SDS
alfa aesar™
H63256MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 214.3
Chemical Name or Material 3-(1-Piperidinylmethyl)indole
Physical Form Solid
Assay 95%
MDL Number MFCD00458412
CAS 5355-42-0
Beilstein 176547
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number NM1303000
Color Off-white to tan
Quantity 1g

Applications
3-(1-Piperidinylmethyl)indole, is used as a pharmaceutical intermediate.

Solubility
Freely soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.