missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(1-Piperidinylmethyl)indole, 95%

Manufacturer:  Alfa Aesar™ H6325606

 View more versions of this product

Catalog No. AAH6325606



Description

Description

Applications
3-(1-Piperidinylmethyl)indole, is used as a pharmaceutical intermediate.

Solubility
Freely soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Specifications

3-(1-Piperidinylmethyl)indole
5g
C14H18N2
176547
HPKXPFRRVVWAPD-UHFFFAOYSA-N
3-(piperidin-1-ylmethyl)-1H-indole
21454
Solid
Tan to White
5355-42-0
MFCD00458412
n-skatylpiperidine, 3-piperidin-1-yl methyl-1h-indole, 3-piperidinomethyl indole, indole, 3-piperidinomethyl, 3-piperidin-1-ylmethyl-1h-indole, chembl57535, 3-piperidinomethyl-1h-indole, 3-1-piperidinylmethyl-1h-indole, 3-piperidylmethyl indole, 3-piperidinomethyl indole
C1CCN(CC1)CC2=CNC3=CC=CC=C32
214.312
214.3
95%

Chemical Identifiers

5355-42-0
214.312
HPKXPFRRVVWAPD-UHFFFAOYSA-N
21454
C1CCN(CC1)CC2=CNC3=CC=CC=C32
C14H18N2
MFCD00458412
n-skatylpiperidine, 3-piperidin-1-yl methyl-1h-indole, 3-piperidinomethyl indole, indole, 3-piperidinomethyl, 3-piperidin-1-ylmethyl-1h-indole, chembl57535, 3-piperidinomethyl-1h-indole, 3-1-piperidinylmethyl-1h-indole, 3-piperidylmethyl indole, 3-piperidinomethyl indole
3-(piperidin-1-ylmethyl)-1H-indole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : NM1303000

SDS Documents