Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(1-Piperazinyl)-1,2-benzisoxazole, 96%

$103.35 - $103.35

Chemical Identifiers

Chemical Name or Material 3-(1-Piperazinyl)-1,2-benzisoxazole
CAS 87691-89-2
Molecular Formula C11H13N3O
Formula Weight 203.25
MDL Number MFCD08060968
Synonym 3-(1-Piperazino)-1,2-benzisoxazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH50746MD

  SDS
alfa aesar™
H50746MD
250mg Each for $103.35
Description & Specifications

Specifications

Formula Weight 203.25
Assay 96%
MDL Number MFCD08060968
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym 3-(1-Piperazino)-1,2-benzisoxazole
Quantity 250mg
Molecular Formula C11H13N3O
CAS 87691-89-2
Melting Point 74° to 76°C
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Chemical Name or Material 3-(1-Piperazinyl)-1,2-benzisoxazole
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.