missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(1-Naphthyl)-L-alanine Hydrochloride, TCI America™

$210.84 - $653.91

Chemical Identifiers

CAS 122745-10-2
Molecular Formula C13H14ClNO2
Molecular Weight (g/mol) 251.71
InChI Key BKQQPCDQZZTLSE-YDALLXLXSA-N
Synonym (S)-alpha-Amino-1-naphthalenepropionic Acid Hydrochloride
PubChem CID 44629853
IUPAC Name (2S)-2-amino-3-naphthalen-1-ylpropanoic acid;hydrochloride
SMILES C1=CC=C2C(=C1)C=CC=C2CC(C(=O)O)N.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N07731G View Documents TCI America
N07731G
1g Each for $210.84
 
N07735G View Documents TCI America
N07735G
5g Each for $653.91
 
Specifications

Chemical Identifiers

122745-10-2
251.71
(S)-alpha-Amino-1-naphthalenepropionic Acid Hydrochloride
(2S)-2-amino-3-naphthalen-1-ylpropanoic acid;hydrochloride
C13H14ClNO2
BKQQPCDQZZTLSE-YDALLXLXSA-N
44629853
C1=CC=C2C(=C1)C=CC=C2CC(C(=O)O)N.Cl

Specifications

Undesignated
122745-10-2
(S)-alpha-Amino-1-naphthalenepropionic Acid Hydrochloride
C1=CC=C2C(=C1)C=CC=C2CC(C(=O)O)N.Cl
251.71
251.71
≥97.0% (HPLC,T)
1g
C13H14ClNO2
BKQQPCDQZZTLSE-YDALLXLXSA-N
(2S)-2-amino-3-naphthalen-1-ylpropanoic acid;hydrochloride
44629853
Crystal-Powder at 20°C
3-(1-Naphthyl)-L-alanine Hydrochloride
Safety and Handling

Safety and Handling

TSCA : No