missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(1-Naphthyl)-L-alanine, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6358603

 View more versions of this product

Catalog No. AAH6358603


Description

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

3-(1-Naphthyl)-L-alanine
C13H13NO2
4352833
Slightly soluble in water.
[NH3+][C@@H](CC1=C2C=CC=CC2=CC=C1)C([O-])=O
215.25
215.25
1 g
55516-54-6
MFCD00066086
l-1-naphthylalanine, 3-1-naphthyl-l-alanine, h-1-nal-oh, s-2-amino-3-naphthalen-1-yl propanoic acid, 3-1-naphthyl-l-ala, unii-9g930ul92r, naphthalen-2-yl-3-alanine, 2s-2-amino-3-naphthalen-1-yl propanoic acid, 3-naphth-1-yl-d-alanine, 1-naphthyl-l-alanine
OFYAYGJCPXRNBL-LBPRGKRZSA-N
(2S)-2-azaniumyl-3-(naphthalen-1-yl)propanoate
2724883
95%
Solid

Chemical Identifiers

55516-54-6
215.25
OFYAYGJCPXRNBL-LBPRGKRZSA-N
2724883
C13H13NO2
MFCD00066086
l-1-naphthylalanine, 3-1-naphthyl-l-alanine, h-1-nal-oh, s-2-amino-3-naphthalen-1-yl propanoic acid, 3-1-naphthyl-l-ala, unii-9g930ul92r, naphthalen-2-yl-3-alanine, 2s-2-amino-3-naphthalen-1-yl propanoic acid, 3-naphth-1-yl-d-alanine, 1-naphthyl-l-alanine
[NH3+][C@@H](CC1=C2C=CC=CC2=CC=C1)C([O-])=O
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only