Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(1-Naphthyl)-D-alanine, 95%

Chemical Identifiers

Chemical Name or Material 3-(1-Naphthyl)-D-alanine
CAS 78306-92-0
Formula Weight 215.25
MDL Number MFCD00038544
Beilstein 4352834
Synonym 3-(1-Naphthyl)-D-Ala-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6640803

  SDS
alfa aesar™
H6640803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6640806

  SDS
alfa aesar™
H6640806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 215.25
Chemical Name or Material 3-(1-Naphthyl)-D-alanine
CAS 78306-92-0
Assay 95%
MDL Number MFCD00038544
Synonym 3-(1-Naphthyl)-D-Ala-OH
Storage Note 1 2° to 8°C
Beilstein 4352834
Quantity 1g
Physical Form Solid

Applications
3-(1-Naphthyl)-D-alanine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in hydrochloric acid.

Notes
Protect from heat. Store away from oxidizing agents. Refrigerate.

Safety and Handling

2° to 8°C