missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(1-Naphthyl)benzeneboronic acid, 98%

$31.61 - $370.11

Chemical Identifiers

CAS 881913-20-8
Molecular Formula C16H13BO2
Molecular Weight (g/mol) 248.088
MDL Number MFCD11045054
InChI Key HFXYUCJLQZCNPD-UHFFFAOYSA-N
Synonym 3-naphthalen-1-yl phenyl boronic acid, 3-naphthalen-1-yl phenylboronic acid, 3-1-naphthyl phenylboronic acid, 3-1-naphthyl benzeneboronic acid, 3-naphthalen-1-yl phenyl boronic acid, boronic acid, 3-1-naphthalenyl phenyl, 3-1-naphthyl phenylboronic acid, 3-1-naphthyl phenyl boronic acid, 3-1-naphthalenyl phenyl-boronic acid, boronic acid, 3-1-naphthalenyl phenyl
PubChem CID 58881077
IUPAC Name (3-naphthalen-1-ylphenyl)boronic acid
SMILES B(C1=CC(=CC=C1)C2=CC=CC3=CC=CC=C32)(O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64019MD View Documents Alfa Aesar™
H64019MD
250mg Each for $31.61
 
AAH6401903 View Documents Alfa Aesar™
H6401903
1g Each for $93.30
 
AAH6401906 View Documents Alfa Aesar™
H6401906
5g Each for $370.11
 
Specifications

Chemical Identifiers

881913-20-8
248.088
HFXYUCJLQZCNPD-UHFFFAOYSA-N
58881077
B(C1=CC(=CC=C1)C2=CC=CC3=CC=CC=C32)(O)O
C16H13BO2
MFCD11045054
3-naphthalen-1-yl phenyl boronic acid, 3-naphthalen-1-yl phenylboronic acid, 3-1-naphthyl phenylboronic acid, 3-1-naphthyl benzeneboronic acid, 3-naphthalen-1-yl phenyl boronic acid, boronic acid, 3-1-naphthalenyl phenyl, 3-1-naphthyl phenylboronic acid, 3-1-naphthyl phenyl boronic acid, 3-1-naphthalenyl phenyl-boronic acid, boronic acid, 3-1-naphthalenyl phenyl
(3-naphthalen-1-ylphenyl)boronic acid

Specifications

White
881913-20-8
MFCD11045054
3-naphthalen-1-yl phenyl boronic acid, 3-naphthalen-1-yl phenylboronic acid, 3-1-naphthyl phenylboronic acid, 3-1-naphthyl benzeneboronic acid, 3-naphthalen-1-yl phenyl boronic acid, boronic acid, 3-1-naphthalenyl phenyl, 3-1-naphthyl phenylboronic acid, 3-1-naphthyl phenyl boronic acid, 3-1-naphthalenyl phenyl-boronic acid, boronic acid, 3-1-naphthalenyl phenyl
B(C1=CC(=CC=C1)C2=CC=CC3=CC=CC=C32)(O)O
248.088
248.09
98%
250mg
C16H13BO2
18694809
HFXYUCJLQZCNPD-UHFFFAOYSA-N
(3-naphthalen-1-ylphenyl)boronic acid
58881077
Powder
3-(1-Naphthyl)benzeneboronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning