missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(1-Naphthyl)benzeneboronic acid, 98%

Manufacturer:  Alfa Aesar™ H64019MD

 View more versions of this product

Catalog No. AAH64019MD



Specifications

Specifications

3-(1-Naphthyl)benzeneboronic acid
250mg
C16H13BO2
18694809
HFXYUCJLQZCNPD-UHFFFAOYSA-N
(3-naphthalen-1-ylphenyl)boronic acid
58881077
Powder
White
881913-20-8
MFCD11045054
3-naphthalen-1-yl phenyl boronic acid, 3-naphthalen-1-yl phenylboronic acid, 3-1-naphthyl phenylboronic acid, 3-1-naphthyl benzeneboronic acid, 3-naphthalen-1-yl phenyl boronic acid, boronic acid, 3-1-naphthalenyl phenyl, 3-1-naphthyl phenylboronic acid, 3-1-naphthyl phenyl boronic acid, 3-1-naphthalenyl phenyl-boronic acid, boronic acid, 3-1-naphthalenyl phenyl
B(C1=CC(=CC=C1)C2=CC=CC3=CC=CC=C32)(O)O
248.088
248.09
98%

Chemical Identifiers

881913-20-8
248.088
HFXYUCJLQZCNPD-UHFFFAOYSA-N
58881077
B(C1=CC(=CC=C1)C2=CC=CC3=CC=CC=C32)(O)O
C16H13BO2
MFCD11045054
3-naphthalen-1-yl phenyl boronic acid, 3-naphthalen-1-yl phenylboronic acid, 3-1-naphthyl phenylboronic acid, 3-1-naphthyl benzeneboronic acid, 3-naphthalen-1-yl phenyl boronic acid, boronic acid, 3-1-naphthalenyl phenyl, 3-1-naphthyl phenylboronic acid, 3-1-naphthyl phenyl boronic acid, 3-1-naphthalenyl phenyl-boronic acid, boronic acid, 3-1-naphthalenyl phenyl
(3-naphthalen-1-ylphenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents