Learn More
Alfa Aesar™ 3-(1-Naphthyl)acrylic acid, 98%
Manufacturer: Alfa Aesar™ A1542230
Description
Applications3-(1-Naphthyl)acrylic acid is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Specifications
3-(1-Naphthyl)acrylic acid | |
250g | |
C13H10O2 | |
1868617 | |
WPXMLUUYWNHQOR-HJWRWDBZSA-N | |
(Z)-3-naphthalen-1-ylprop-2-enoic acid | |
12158295 | |
98% |
210°C to 212°C | |
13026-12-5 | |
MFCD00014317 | |
naphthalene-1-acrylic acid, z-3-1-naphthyl acrylic acid, z-3-naphthalen-1-yl-acrylic acid, z-3-naphthalen-1-yl acrylic acid, 2z-3-naphthalen-1-yl prop-2-enoic acid | |
C1=CC=C2C(=C1)C=CC=C2C=CC(=O)O | |
198.221 | |
198.22 |
Chemical Identifiers
13026-12-5 | |
198.221 | |
WPXMLUUYWNHQOR-HJWRWDBZSA-N | |
12158295 | |
C1=CC=C2C(=C1)C=CC=C2C=CC(=O)O |
C13H10O2 | |
MFCD00014317 | |
naphthalene-1-acrylic acid, z-3-1-naphthyl acrylic acid, z-3-naphthalen-1-yl-acrylic acid, z-3-naphthalen-1-yl acrylic acid, 2z-3-naphthalen-1-yl prop-2-enoic acid | |
(Z)-3-naphthalen-1-ylprop-2-enoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning