missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(1-Methyl-1H-pyrazol-5-yl)benzonitrile, 97%, Maybridge

Chemical Identifiers

CAS 179055-95-9
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C11H9N3
Formula Weight 183.21
MDL Number MFCD09702401
Chemical Name or Material 3-(1-Methyl-1H-pyrazol-5-yl)benzonitrile
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC60516DE SDS Fisher Scientific Company
CC60516DE
Amber Glass Bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

179055-95-9
100.0
C11H9N3
MFCD09702401
97%
96.5
97%
183.21
3-(1-Methyl-1H-pyrazol-5-yl)benzonitrile

Specifications

5g
96.5
Amber Glass Bottle
C11H9N3
MFCD09702401
97%
179055-95-9
100.0
97%
183.21
3-(1-Methyl-1H-pyrazol-5-yl)benzonitrile

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning