missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(1-methyl-1H-pyrazol-3-yl)aniline, 97+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC60614CB
Specifications
3-(1-Methyl-1H-pyrazol-3-yl)aniline | |
95% | |
C10H11N3 | |
3-1-methyl-1h-pyrazol-3-yl aniline, 3-1-methylpyrazol-3-yl aniline, 3-1-methyl-1h-pyrazol-3-yl-phenylamine, benzenamine, 3-1-methyl-1h-pyrazol-3-yl, 3-1-methylpyrazol-3-yl phenylamine, 3-1-methyl-1h-pyrazol-3-yl phenylamine, 3-3-aminophenyl-1-methyl-1h-pyrazole | |
CN1C=CC(=N1)C2=CC(=CC=C2)N | |
173.219 | |
173.22 |
175202-37-6 | |
Amber Glass Bottle | |
250mg | |
ZRXOZSROZJGRRH-UHFFFAOYSA-N | |
3-(1-methylpyrazol-3-yl)aniline | |
2735396 | |
97% |
Chemical Identifiers
175202-37-6 | |
173.219 | |
3-1-methyl-1h-pyrazol-3-yl aniline, 3-1-methylpyrazol-3-yl aniline, 3-1-methyl-1h-pyrazol-3-yl-phenylamine, benzenamine, 3-1-methyl-1h-pyrazol-3-yl, 3-1-methylpyrazol-3-yl phenylamine, 3-1-methyl-1h-pyrazol-3-yl phenylamine, 3-3-aminophenyl-1-methyl-1h-pyrazole | |
3-(1-methylpyrazol-3-yl)aniline |
C10H11N3 | |
ZRXOZSROZJGRRH-UHFFFAOYSA-N | |
2735396 | |
CN1C=CC(=N1)C2=CC(=CC=C2)N |
Safety and Handling
GHS Signal Word: Warning