Please login to your online account to display your discounted pricing

3-(1-methyl-1H-pyrazol-3-yl)aniline, 97+%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC60614DE

 View more versions of this product

Catalog No. CC60614DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 173.22
Quantity 5g
Assay 97%
Chemical Name or Material 3-(1-Methyl-1H-pyrazol-3-yl)aniline
Packaging Amber Glass Bottle
Assay Percent Range 95%
CAS 175202-37-6
Molecular Formula C10H11N3
Health Hazard 1 GHS Signal Word: Warning

Specifications

Chemical Name or Material 3-(1-Methyl-1H-pyrazol-3-yl)aniline
Assay 97%
Assay Percent Range 95%
Molecular Formula C10H11N3
Formula Weight 173.22
Accessories

GHS Signal Word: Warning