missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(1-Cyanoethyl)benzoic acid, 98%

Manufacturer:  Alfa Aesar™ H6053703

 View more versions of this product

Catalog No. AAH6053703



Specifications

Specifications

3-(1-Cyanoethyl)benzoic acid
1g
C10H9NO2
3-1-cyanoethyl benzoic acid, m-1-cyanoethyl benzoic acid, benzoic acid, 3-1-cyanoethyl, zlchem 504, acmc-1b01z, 3-1-cyanethyl benzoesaeure, 3-1-cyanoethyl benzoicacid, 3-l-cyanoethyl benzoic acid, 3-1-cyanethyl benzoic acid, meta-1-cyanoethyl benzoic acid
CC(C#N)C1=CC=CC(=C1)C(=O)O
175.187
CHEBI:85131
98%
145°C to 148°C
5537-71-3
MFCD00002520
IRYIYPWRXROPSX-UHFFFAOYSA-N
3-(1-cyanoethyl)benzoic acid
98370
175.18

Chemical Identifiers

5537-71-3
175.187
IRYIYPWRXROPSX-UHFFFAOYSA-N
98370
3-(1-cyanoethyl)benzoic acid
C10H9NO2
MFCD00002520
3-1-cyanoethyl benzoic acid, m-1-cyanoethyl benzoic acid, benzoic acid, 3-1-cyanoethyl, zlchem 504, acmc-1b01z, 3-1-cyanethyl benzoesaeure, 3-1-cyanoethyl benzoicacid, 3-l-cyanoethyl benzoic acid, 3-1-cyanethyl benzoic acid, meta-1-cyanoethyl benzoic acid
CHEBI:85131
CC(C#N)C1=CC=CC(=C1)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H301-H318-H317
Toxic if swallowed.
Causes serious eye damage.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

SDS Documents