missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(1-Cyanoethyl)benzoic acid, 98%

$19.14 - $82.92

Chemical Identifiers

CAS 5537-71-3
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD00002520
InChI Key IRYIYPWRXROPSX-UHFFFAOYSA-N
Synonym 3-1-cyanethyl benzoesaeure, 3-1-cyanethyl benzoic acid, 3-1-cyanoethyl benzoic acid, 3-1-cyanoethyl benzoicacid, 3-l-cyanoethyl benzoic acid, acmc-1b01z, benzoic acid, 3-1-cyanoethyl, m-1-cyanoethyl benzoic acid, meta-1-cyanoethyl benzoic acid, zlchem 504
PubChem CID 98370
ChEBI CHEBI:85131
IUPAC Name 3-(1-cyanoethyl)benzoic acid
SMILES CC(C#N)C1=CC=CC(=C1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6053703
View Documents
Alfa Aesar™
H6053703
1g Each for $19.14
 
AAH6053706
View Documents
Alfa Aesar™
H6053706
5g Each for $82.92
 
Specifications

Chemical Identifiers

5537-71-3
175.187
IRYIYPWRXROPSX-UHFFFAOYSA-N
98370
3-(1-cyanoethyl)benzoic acid
C10H9NO2
MFCD00002520
3-1-cyanethyl benzoesaeure, 3-1-cyanethyl benzoic acid, 3-1-cyanoethyl benzoic acid, 3-1-cyanoethyl benzoicacid, 3-l-cyanoethyl benzoic acid, acmc-1b01z, benzoic acid, 3-1-cyanoethyl, m-1-cyanoethyl benzoic acid, meta-1-cyanoethyl benzoic acid, zlchem 504
CHEBI:85131
CC(C#N)C1=CC=CC(=C1)C(=O)O

Specifications

145°C to 148°C
5537-71-3
MFCD00002520
IRYIYPWRXROPSX-UHFFFAOYSA-N
3-(1-cyanoethyl)benzoic acid
98370
175.18
3-(1-Cyanoethyl)benzoic acid
1g
C10H9NO2
3-1-cyanethyl benzoesaeure, 3-1-cyanethyl benzoic acid, 3-1-cyanoethyl benzoic acid, 3-1-cyanoethyl benzoicacid, 3-l-cyanoethyl benzoic acid, acmc-1b01z, benzoic acid, 3-1-cyanoethyl, m-1-cyanoethyl benzoic acid, meta-1-cyanoethyl benzoic acid, zlchem 504
CC(C#N)C1=CC=CC(=C1)C(=O)O
175.187
CHEBI:85131
98%
Safety and Handling

Safety and Handling

GHS H Statement
H301-H318-H317
Toxic if swallowed.
Causes serious eye damage.
May cause an allergic skin reaction.

P264b-P280i-P305+P351+P338

H319

EINECSNumber : 226-897-0

TSCA : No

Recommended Storage : Ambient temperatures