missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(1-Cyanoethyl)benzoic acid, 98%

Manufacturer:  Alfa Aesar™ H6053706

 View more versions of this product

Catalog No. AAH6053706



Specifications

Specifications

3-(1-Cyanoethyl)benzoic acid
5g
C10H9NO2
3-1-cyanethyl benzoesaeure, 3-1-cyanethyl benzoic acid, 3-1-cyanoethyl benzoic acid, 3-1-cyanoethyl benzoicacid, 3-l-cyanoethyl benzoic acid, acmc-1b01z, benzoic acid, 3-1-cyanoethyl, m-1-cyanoethyl benzoic acid, meta-1-cyanoethyl benzoic acid, zlchem 504
CC(C#N)C1=CC=CC(=C1)C(=O)O
175.187
CHEBI:85131
98%
145°C to 148°C
5537-71-3
MFCD00002520
IRYIYPWRXROPSX-UHFFFAOYSA-N
3-(1-cyanoethyl)benzoic acid
98370
175.18

Chemical Identifiers

5537-71-3
175.187
IRYIYPWRXROPSX-UHFFFAOYSA-N
98370
3-(1-cyanoethyl)benzoic acid
C10H9NO2
MFCD00002520
3-1-cyanethyl benzoesaeure, 3-1-cyanethyl benzoic acid, 3-1-cyanoethyl benzoic acid, 3-1-cyanoethyl benzoicacid, 3-l-cyanoethyl benzoic acid, acmc-1b01z, benzoic acid, 3-1-cyanoethyl, m-1-cyanoethyl benzoic acid, meta-1-cyanoethyl benzoic acid, zlchem 504
CHEBI:85131
CC(C#N)C1=CC=CC(=C1)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H301-H318-H317
Toxic if swallowed.
Causes serious eye damage.
May cause an allergic skin reaction.

P264b-P280i-P305+P351+P338

H319

EINECSNumber : 226-897-0

TSCA : No

Recommended Storage : Ambient temperatures

Documents