missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[3-(1,3-Thiazol-2-yl)phenyl]methylamine, 97%, Maybridge

Chemical Identifiers

CAS 672324-88-8
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C10H10N2S
Formula Weight 190.27
MDL Number MFCD08741424
Chemical Name or Material [3-(1, 3-Thiazol-2-yl)phenyl]methylamine
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC36513DA SDS Fisher Scientific Company
CC36513DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC36513CB SDS Fisher Scientific Company
CC36513CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

672324-88-8
100.0
C10H10N2S
MFCD08741424
97%
96.5
95%
190.27
[3-(1, 3-Thiazol-2-yl)phenyl]methylamine

Specifications

1g
96.5
Amber Glass Bottle
C10H10N2S
MFCD08741424
97%
672324-88-8
100.0
95%
190.27
[3-(1, 3-Thiazol-2-yl)phenyl]methylamine

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger