Please login to your online account to display your discounted pricing

[3-(1,3-Thiazol-2-yl)phenyl]methylamine, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC36513CB

 View more versions of this product

Catalog No. CC36513CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 190.27
Quantity 250mg
Assay 97%
CAS Max % 100.0
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 672324-88-8
MDL Number MFCD08741424
CAS Min % 96.5
Packaging Amber Glass Bottle
Chemical Name or Material [3-(1, 3-Thiazol-2-yl)phenyl]methylamine
Molecular Formula C10H10N2S
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Assay Percent Range 95%

Specifications

Chemical Name or Material [3-(1, 3-Thiazol-2-yl)phenyl]methylamine
CAS 672324-88-8
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 95%
Molecular Formula C10H10N2S
Formula Weight 190.27
MDL Number MFCD08741424
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents