missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(1,2,3,6-Tetrahydro-4-pyridyl)indole, 95%

$72.47 - $791.11

Chemical Identifiers

CAS 65347-55-9
Molecular Formula C13H14N2
Molecular Weight (g/mol) 198.269
MDL Number MFCD03695471
InChI Key CIRSPTXGPFAXRE-UHFFFAOYSA-N
Synonym 3-1,2,3,6-tetrahydropyridin-4-yl-1h-indole, chembl27811, 3-1,2,3,6-tetrahydro-4-pyridinyl-1h-indole, 3-1,2,3,6-tetrahydro-pyridin-4-yl-1h-indole, 3-4-1,2,5,6-tetrahydropyridyl indole, thpindole, d02mvr, 3-1,2,3,6-tetrahydropyridin-4-yl indole, 3-1,2,3,6-tetrahydro-4-pyridinyl 1h-indol, 3-1,2,3,6-tetrahydro-4-pyridinyl 1h-indole
PubChem CID 2761023
IUPAC Name 3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-indole
SMILES C1CNCC=C1C2=CNC3=CC=CC=C32
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6382103 View Documents Alfa Aesar™
H6382103
1g Each for $174.26
 
AAH6382106 View Documents Alfa Aesar™
H6382106
5g Each for $791.11
 
AAH63821MD View Documents Alfa Aesar™
H63821MD
250mg Each for $72.47
 
Specifications

Chemical Identifiers

65347-55-9
198.269
CIRSPTXGPFAXRE-UHFFFAOYSA-N
2761023
C1CNCC=C1C2=CNC3=CC=CC=C32
C13H14N2
MFCD03695471
3-1,2,3,6-tetrahydropyridin-4-yl-1h-indole, chembl27811, 3-1,2,3,6-tetrahydro-4-pyridinyl-1h-indole, 3-1,2,3,6-tetrahydro-pyridin-4-yl-1h-indole, 3-4-1,2,5,6-tetrahydropyridyl indole, thpindole, d02mvr, 3-1,2,3,6-tetrahydropyridin-4-yl indole, 3-1,2,3,6-tetrahydro-4-pyridinyl 1h-indol, 3-1,2,3,6-tetrahydro-4-pyridinyl 1h-indole
3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-indole

Specifications

1g
C13H14N2
958099
CIRSPTXGPFAXRE-UHFFFAOYSA-N
3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-indole
2761023
Solid
3-(1,2,3,6-Tetrahydro-4-pyridyl)indole
65347-55-9
MFCD03695471
3-1,2,3,6-tetrahydropyridin-4-yl-1h-indole, chembl27811, 3-1,2,3,6-tetrahydro-4-pyridinyl-1h-indole, 3-1,2,3,6-tetrahydro-pyridin-4-yl-1h-indole, 3-4-1,2,5,6-tetrahydropyridyl indole, thpindole, d02mvr, 3-1,2,3,6-tetrahydropyridin-4-yl indole, 3-1,2,3,6-tetrahydro-4-pyridinyl 1h-indol, 3-1,2,3,6-tetrahydro-4-pyridinyl 1h-indole
C1CNCC=C1C2=CNC3=CC=CC=C32
198.269
198.26
95%