Please login to your online account to display your discounted pricing

3-(1,1,2,3,3,3-Hexafluoropropyl)adamantane-1-carboxylic acid, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 3-(1,1,2,3,3,3-Hexafluoropropyl)adamantane-1-carboxylic acid
CAS 86301-98-6
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C14H16F6O2
Formula Weight 330.27
MDL Number MFCD00193108
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RJC02875CB

 
fisher scientific company
RJC02875CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 330.27
Quantity 250mg
Assay 97%
CAS Min % 97.0
Assay Percent Range 97%
MDL Number MFCD00193108
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C14H16F6O2
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Packaging Amber Glass Bottle
Chemical Name or Material 3-(1,1,2,3,3,3-Hexafluoropropyl)adamantane-1-carboxylic acid
CAS 86301-98-6