Please login to your online account to display your discounted pricing

2xYT Broth (Powder/Molecular Genetics), Fisher BioReagents


Manufacturer: fisher scientific company  BP2467500

 View more versions of this product

Catalog No. BP2467-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Specifications

Packaging Poly bottle
Quantity 500g
Storage Requirements <4°C
For Use With (Application) Molecular Genetics
Formulation 31g/L
Format Powder
Additive 2xyt Broth