Please login to your online account to display your discounted pricing

2xYT Broth (Powder/Molecular Genetics), Fisher BioReagents


Manufacturer: fisher scientific company  BP2467100

 View more versions of this product

Catalog No. BP2467-100

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Specifications

Additive 2xyt Broth
For Use With (Application) Molecular Genetics
Format Powder
Formulation 31g/L
Packaging Poly bottle
Quantity 100g
Storage Requirements <4°C