Please login to your online account to display your discounted pricing

2xYT Agar (Powder/Molecular Genetics), Fisher BioReagents


Manufacturer: fisher scientific company  BP2466500

 View more versions of this product

Catalog No. BP2466-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Specifications

Additive 2xyt Agar
For Use With (Application) Molecular Genetics
Format Powder
Packaging Poly bottle
Quantity 500g