Please login to your online account to display your discounted pricing

2xYT Agar (Powder/Molecular Genetics), Fisher BioReagents


Manufacturer: fisher scientific company  BP24662

 View more versions of this product

Catalog No. BP2466-2

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Specifications

Format Powder
Additive 2xyt Agar
Quantity 2000g
For Use With (Application) Molecular Genetics
Packaging Poly bottle