Learn More
(2S)-(+)-Glycidyl tosylate, 99+%, Thermo Scientific™
Chemical Identifiers
CAS | 70987-78-9 |
---|---|
Molecular Formula | C10H12O4S |
Molecular Weight (g/mol) | 228.27 |
MDL Number | MFCD00064489 |
InChI Key | NOQXXYIGRPAZJC-VIFPVBQESA-N |
Synonym | s-glycidyl tosylate, 2s-+-glycidyl tosylate, s-+-glycidyl tosylate, +-glycidyl tosylate, s-oxiran-2-ylmethyl 4-methylbenzenesulfonate, 2s-oxiran-2-ylmethyl 4-methylbenzenesulfonate, ccris 6400, s-glycidyltosylate, 2s-glycidyl tosylate, oxiranemethanol, 4-methylbenzenesulfonate, s |
PubChem CID | 153296 |
IUPAC Name | [(2S)-oxiran-2-yl]methyl 4-methylbenzenesulfonate |
SMILES | CC1=CC=C(C=C1)S(=O)(=O)OCC2CO2 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC264580050
|
Thermo Scientific Chemicals
264580050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC264580250
|
Thermo Scientific Chemicals
264580250 |
25g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
70987-78-9 | |
228.27 | |
NOQXXYIGRPAZJC-VIFPVBQESA-N | |
153296 | |
CC1=CC=C(C=C1)S(=O)(=O)OCC2CO2 |
C10H12O4S | |
MFCD00064489 | |
s-glycidyl tosylate, 2s-+-glycidyl tosylate, s-+-glycidyl tosylate, +-glycidyl tosylate, s-oxiran-2-ylmethyl 4-methylbenzenesulfonate, 2s-oxiran-2-ylmethyl 4-methylbenzenesulfonate, ccris 6400, s-glycidyltosylate, 2s-glycidyl tosylate, oxiranemethanol, 4-methylbenzenesulfonate, s | |
[(2S)-oxiran-2-yl]methyl 4-methylbenzenesulfonate |
Specifications
70987-78-9 | |
100.0 | |
Authentic | |
Glass bottle | |
MFCD00064489 | |
+ 17.00 (20.00°C c=2.5,CHCl3) | |
s-glycidyl tosylate, 2s-+-glycidyl tosylate, s-+-glycidyl tosylate, +-glycidyl tosylate, s-oxiran-2-ylmethyl 4-methylbenzenesulfonate, 2s-oxiran-2-ylmethyl 4-methylbenzenesulfonate, ccris 6400, s-glycidyltosylate, 2s-glycidyl tosylate, oxiranemethanol, 4-methylbenzenesulfonate, s | |
NOQXXYIGRPAZJC-VIFPVBQESA-N | |
[(2S)-oxiran-2-yl]methyl 4-methylbenzenesulfonate | |
153296 | |
Fluffy Powder | |
(2S)-(+)-Glycidyl tosylate |
99.0 | |
>110°C | |
99% min. (GC) | |
C10H12O4S | |
5g | |
+ 17 | |
Solubility in water: decomposes. Other solubilities: soluble in chloroform | |
CC1=CC=C(C=C1)S(=O)(=O)OCC2CO2 | |
228.27 | |
228.27 | |
99+% |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.
Causes serious eye damage.
Suspected of causing genetic defects.
May cause cancer.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF exposed or concerned: Get medi
GHS Signal Word: Danger
RUO â Research Use Only