missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2S)-(+)-Glycidyl tosylate, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 264580010

 View more versions of this product

Catalog No. AC264580010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

(2S)-(+)-Glycidyl tosylate
Authentic
+ 17.00 (20.00°C c=2.5,CHCl3)
White
1g
99.0
99% min. (GC)
MFCD00064489
Solubility in water: decomposes. Other solubilities: soluble in chloroform
CC1=CC=C(C=C1)S(=O)(=O)OCC2CO2
228.27
228.27
99+%
>110°C
Glass bottle
+ 17
45°C to 48°C
70987-78-9
100.0
C10H12O4S
+-glycidyl tosylate, 2s-+-glycidyl tosylate, 2s-glycidyl tosylate, 2s-oxiran-2-ylmethyl 4-methylbenzenesulfonate, ccris 6400, oxiranemethanol, 4-methylbenzenesulfonate, s, s-+-glycidyl tosylate, s-glycidyl tosylate, s-glycidyltosylate, s-oxiran-2-ylmethyl 4-methylbenzenesulfonate
NOQXXYIGRPAZJC-VIFPVBQESA-N
[(2S)-oxiran-2-yl]methyl 4-methylbenzenesulfonate
153296
Fluffy Powder

Chemical Identifiers

70987-78-9
228.27
NOQXXYIGRPAZJC-VIFPVBQESA-N
153296
CC1=CC=C(C=C1)S(=O)(=O)OCC2CO2
C10H12O4S
MFCD00064489
+-glycidyl tosylate, 2s-+-glycidyl tosylate, 2s-glycidyl tosylate, 2s-oxiran-2-ylmethyl 4-methylbenzenesulfonate, ccris 6400, oxiranemethanol, 4-methylbenzenesulfonate, s, s-+-glycidyl tosylate, s-glycidyl tosylate, s-glycidyltosylate, s-oxiran-2-ylmethyl 4-methylbenzenesulfonate
[(2S)-oxiran-2-yl]methyl 4-methylbenzenesulfonate
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.
Causes serious eye damage.
Suspected of causing genetic defects.
May cause cancer.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF exposed or concerned: Get medi

GHS Signal Word: Danger

SDS Documents