missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (2S)-(+)-Glycidyl p-toluenesulfonate, 99%

$43.39 - $591.45

Chemical Identifiers

CAS 70987-78-9
Molecular Formula C10H12O4S
Molecular Weight (g/mol) 228.262
MDL Number MFCD00064489
InChI Key NOQXXYIGRPAZJC-VIFPVBQESA-N
Synonym +-glycidyl tosylate
PubChem CID 153296
IUPAC Name [(2S)-oxiran-2-yl]methyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCC2CO2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5600003 View Documents Alfa Aesar™
H5600003
1g Each for $43.39
 
AAH5600006 View Documents Alfa Aesar™
H5600006
5g Each for $154.95
 
AAH5600014 View Documents Alfa Aesar™
H5600014
25g Each for $591.45
 
Specifications

Chemical Identifiers

70987-78-9
228.262
NOQXXYIGRPAZJC-VIFPVBQESA-N
153296
CC1=CC=C(C=C1)S(=O)(=O)OCC2CO2
C10H12O4S
MFCD00064489
+-glycidyl tosylate
[(2S)-oxiran-2-yl]methyl 4-methylbenzenesulfonate

Specifications

113°C (235°F)
1g
C10H12O4S
UN3077
+-glycidyl tosylate
CC1=CC=C(C=C1)S(=O)(=O)OCC2CO2
228.262
228.26
(2S)-(+)-Glycidyl p-toluenesulfonate
46°C to 49°C
70987-78-9
MFCD00064489
3592143
NOQXXYIGRPAZJC-VIFPVBQESA-N
[(2S)-oxiran-2-yl]methyl 4-methylbenzenesulfonate
153296
99%
Safety and Handling

Safety and Handling

GHS H Statement
H350-H341-H318-H317
May cause cancer.
Suspected of causing genetic defects.
Causes serious eye damage.
May cause an allergic skin reaction.

P201-P202-P261-P272-P280g-P281-P302+P352-P305+P351+P338-P308+P313-P310-P333+P313-P363-P501c

H317-H318-H341-H350

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 417-210-7

RTECSNumber : RR0510050

TSCA : No

Recommended Storage : Ambient temperatures