Please login to your online account to display your discounted pricing

(2S-cis)-(+)-2,3-Dihydro-3-hydroxy-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one, 98%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  334220050

 View more versions of this product

Catalog No. AC334220050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Powder
Formula Weight 301.36
Quantity 5g
Color White to slightly beige
EINECS Number 255-799-0
TSCA TSCA
Melting Point 203° to 207°C
Specific Rotation Condition +110° (114°C )
CAS Max % 100.0
Specific Rotation +110° to +114° (20°C, 589nm) (c=0.5, CH3OH)
MDL Number MFCD01321316
CAS Min % 97.5
Chemical Name or Material (2S-cis)-(+)-2, 3-Dihydro-3-hydroxy-2-(4-methoxyphenyl)-1, 5-benzothiazepin-4(5H)-one
Infrared Spectrum Authentic
Name Note 98%
CAS 42399-49-5
Molecular Formula C16H15NO3S
Assay Percent Range 98%

Specifications

Chemical Name or Material (2S-cis)-(+)-2, 3-Dihydro-3-hydroxy-2-(4-methoxyphenyl)-1, 5-benzothiazepin-4(5H)-one
Name Note 98%
CAS 42399-49-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C16H15NO3S
Formula Weight 301.36
MDL Number MFCD01321316